DL-Carnitine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16589-14 A16589-22
CAAAA16589-14EA 48.03 CAD
CAAAA16589-14 CAAAA16589-22
DL-Carnitine hydrochloride 98+%
DL-Carnitine hydrochloride

Purity: 98+% CAS No.: 461-05-2 Molecular Formula: C7H15NO3.HCl Formula Weight: 197.66 Melting Point: 196-200° Beilstein Reference: 4163212 Merck Reference: 14,1849 MDL No.: MFCD00011904

Formula: C₇H₁₆ClNO₃
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00011904
CAS Number: 461-05-2
EINECS: 207-309-1
Merck Index: 14,1849

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR