(+)-Dibenzoyl-D-tartaric acid monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2S,3S)-2,3-Bis(benzoyloxy)succinic acid hydrate
A16503-14
A16503-22
CAAAA16503-14EA
79.63
CAD
CAAAA16503-14
CAAAA16503-22
(+)-Dibenzoyl-D-tartaric acid monohydrate 99%
(+)-Dibenzoyl-D-tartaric acid monohydrate
Purity: 99%
CAS No.: 80822-15-7
Molecular Formula: C18H14O8.H2O
Formula Weight: 376.33 (358.31anhy)
Melting Point: 87-91°
Beilstein Reference: 6121616
Formula:
C₁₈H₁₄O₈·1H₂O MW: 376.33 g/mol (358.31 anhy) Melting Pt: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150722 CAS Number: 80822-15-7 EINECS: 241-097-1 |
Specification Test Results
Optical rotation | +109° (c=2 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...