Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16223-03
A16223-06
A16223-14
CAAAA16223-03EA
73.94
CAD
CAAAA16223-03
CAAAA16223-06
CAAAA16223-14
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Benzo-18-crown-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Purity: 97%
CAS No.: 14098-24-9
Molecular Formula: C16H24O6
Formula Weight: 312.37
Melting Point: 43-45°
Beilstein Reference: 1626123
Formula:
C₁₆H₂₄O₆ Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062741 CAS Number: 14098-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...