(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning

A16181-14 A16181-22 A16181-36
CAAAAA16181-14EA 56.88 CAD
CAAAAA16181-14 CAAAA16181-22 CAAAA16181-36
(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%
(-)-Dibenzoyl-L-tartaric acid

Purity: 99% CAS No.: 2743-38-6 Molecular Formula: C18H14O8 Formula Weight: 358.31 Melting Point: 154-156° MDL No.: MFCD00003074

Formula: C₁₈H₁₄O₈
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00003074
CAS Number: 2743-38-6
EINECS: 220-374-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR