(-)-Dibenzoyl-L-tartaric acid monohydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16180-0E A16180-22 A16180-36
CAAAA16180-0EEA 649.38 CAD
CAAAA16180-0E CAAAA16180-22 CAAAA16180-36
(-)-Dibenzoyl-L-tartaric acid monohydrate 98+%
(-)-Dibenzoyl-L-tartaric acid monohydrate

Purity: 98+% CAS No.: 62708-56-9 Molecular Formula: C18H14O8.H2O Formula Weight: 376.33 (358.31anhy) Melting Point: 90-92° MDL No.: MFCD00149119

Formula: C₁₈H₁₄O₈·1H₂O
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00149119
CAS Number: 62708-56-9
EINECS: 613-072-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR