Triethylenetetramine mixture of isomers 60% balance branched and cyclic triethylenetetramines, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16128-0F
A16128-AP
CAAAA16128-0FEA
260.7
CAD
CAAAA16128-0F
CAAAA16128-AP
Triethylenetetramine mixture of isomers 60% balance branched and cyclic triethylenetetramines, Technical Grade
Trientine
Catalyzes the hydrolysis of N-terminal peptide bonds
Purity: 60%
CAS No.: 112-24-3
Molecular Formula: C6H18N4
Formula Weight: 146.24
Flash Point: 122°(252°F)
Refractive Index: 1.4980
Odor: Amine-like
Melting Point: ca -35°
Boiling Point: ca 267°
Beilstein Reference: 605448
Merck Reference: 14,9663
UN No.: 2259
MDL No.: MFCD00008169
Formula:
C₆H₁₈N₄ Boiling Pt: ∼ 267 °C Melting Pt: ∼ 35 °C Density: 0.982 Flash Pt: 122 °C (252 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008169 CAS Number: 112-24-3 EINECS: 203-950-6 UN: 2259 ADR: 8,II Merck Index: 14,9663 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...