Acetamidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16091-14
A16091-22
A16091-36
CAAAAA16091-14EA
42.82
CAD
CAAAAA16091-14
CAAAA16091-22
CAAAA16091-36
Acetamidine hydrochloride 97%
Acetamidine hydrochloride
Purity: 97%
CAS No.: 124-42-5
Molecular Formula: C2H6N2.HCl
Formula Weight: 94.54
Melting Point: 168-175°
Beilstein Reference: 3591762
Merck Reference: 14,44
MDL No.: MFCD00013016
Formula:
C₂H₇ClN₂ Melting Pt: 168…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013016 CAS Number: 124-42-5 EINECS: 204-700-9 Merck Index: 14,44 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...