Glyoxylic acid monohydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Dihydroxyacetic acid, Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate

A16058-14 A16058-22
CAAAA16058-14EA 81.37 CAD
CAAAA16058-14 CAAAA16058-22
Glyoxylic acid monohydrate 97%
Glyoxylic acid monohydrate

Fieser: 5,320 7,162 9,228


Purity: 97% CAS No.: 563-96-2 Molecular Formula: OHCCO2H.H2O Formula Weight: 92.06 (74.04anhy) Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 100° Beilstein Reference: 969535 Merck Reference: 14,4511 UN No.: 3261 MDL No.: MFCD00149497

Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 48…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00149497
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: "8",III
Merck Index: 14,4511

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR