Allyltriphenylphosphonium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Allyltriphenylphosphonium Chloride

A16012-14 A16012-22 A16012-36
CAAAA16012-14EA 86.58 CAD
CAAAA16012-14 CAAAA16012-22 CAAAA16012-36
Allyltriphenylphosphonium chloride 99%
Allyltriphenylphosphonium chloride

Purity: 99% CAS No.: 18480-23-4 Molecular Formula: C21H20ClP Formula Weight: 338.82 Melting Point: 230-232° MDL No.: MFCD00031542

Formula: C₂₁H₂₀ClP
MW: 338.82 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00031542
CAS Number: 18480-23-4
EINECS: 242-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR