Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo-15-crown-5

A16011-03 A16011-06
CAAAA16011-03EA 73.63 CAD
CAAAA16011-03 CAAAA16011-06
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
Benzo-15-crown-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecane)

Purity: 98% CAS No.: 14098-44-3 Molecular Formula: C14H20O5 Formula Weight: 268.31 Melting Point: 78-82° Beilstein Reference: 1624106

Formula: C₁₄H₂₀O₅
MW: 268.31 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00005945
CAS Number: 14098-44-3
EINECS: 237-947-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR