N-Methylmorpholine N-oxide monohydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15996-14
A15996-22
A15996-09
A15996-18
CAAAAA15996-14EA
125.77
CAD
CAAAAA15996-14
CAAAAA15996-22
CAAAA15996-09
CAAAA15996-18
N-Methylmorpholine N-oxide monohydrate 98+%
N-Methylmorpholine N-oxide monohydrate
Fieser: 1,690 3,204 7,244 13,222 19,216 20,245 21,269
Purity: 98+%
CAS No.: 70187-32-5
Molecular Formula: C5H11NO2.H2O
Formula Weight: 135.17 (117.15anhy)
Melting Point: 73-76°
Beilstein Reference: 5449441
Formula:
C₅H₁₁NO₂·1H₂O Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149388 CAS Number: 70187-32-5 EINECS: 231-391-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...