L-Methionine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Met-OMe.HCl, H-Met-OMe·HCl

A15952-06 A15952-14 A15952-22
CAAAA15952-06EA 59.72 CAD
CAAAA15952-06 CAAAA15952-14 CAAAA15952-22
L-Methionine methyl ester hydrochloride 99%
L-Methionine methyl ester hydrochloride

Purity: 99% CAS No.: 2491-18-1 Molecular Formula: C6H13NO2S.HCl Formula Weight: 199.70 Storage Temperature: -30°C to -10°C Melting Point: 151-152° Beilstein Reference: 3913214 Optical Rotation: +26° (c=5 in water) MDL No.: MFCD00012491

Formula: C₆H₁₃NO₂S·HCl
MW: 199.7 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Freezer
MDL Number: MFCD00012491
CAS Number: 2491-18-1
EINECS: 219-651-9

Order Now

Specification Test Results

Optical rotation +26° (c=5 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR