N-Cbz-glycine(Z-Gly-OH) 98+%
Supplier: Thermo Scientific Chemicals
A15928-14
A15928-22
CAAAA15928-14EA
71.57
CAD
CAAAA15928-14
CAAAA15928-22
CAAAA15928-36
N-Cbz-glycine(Z-Gly-OH) 98+%
N-Cbz-Glycine (Z-Gly-OH)
Purity: 98+%
CAS No.: 1138-80-3
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Storage Temperature: 2°C to 8°C
Melting Point: 119-122°
Beilstein Reference: 526877
MDL No.: MFCD00002691
Formula:
C₁₀H₁₁NO₄ Melting Pt: 119…122 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002691 CAS Number: 1138-80-3 EINECS: 214-516-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...