Brij® 35
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
A15809-0B
A15809-0I
A15809-30
CAAAA15809-0BEA
127.33
CAD
CAAAA15809-0B
CAAAA15809-0I
CAAAA15809-30
Brij® 35
Brij® 35
For use in Stein-Moore chromatography
CAS No.: 9002-92-0
Molecular Formula: C58H118O24
Formula Weight: 1199.57
Physical Form: Waxy solid
Appearance: White
Melting Point: ca 35-40°
MDL No.: MFCD00080891
For use in Stein-Moore chromatography
Formula:
C₅₈H₁₁₈O₂₄ MW: 1199.56 g/mol Melting Pt: ∼ 35…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080891 CAS Number: 9002-92-0 EINECS: 500-002-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...