Sodium hydroxymethanesulphinate, hydrate tech. 85% (dry weight) cont. up to 5% sodium sulfite, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium formaldehydesulfoxylate, Sodium formaldehydesulphoxylate

A15731-0E A15731-36
CAAAA15731-0EEA 218.04 CAD
CAAAA15731-0E CAAAA15731-36
Sodium hydroxymethanesulphinate, hydrate tech. 85% (dry weight) cont. up to 5% sodium sulfite, Technical Grade
Sodium hydroxymethanesulphinate

Purity: 85% CAS No.: 149-44-0 Molecular Formula: HOCH2SO2Na.xH2O Formula Weight: 118.09(anhy) Melting Point: 64-68° Beilstein Reference: 3630814 Merck Reference: 14,8620 MDL No.: MFCD00150598

Formula: CH₃NaO₃S·H₂O
MW: 118.09 g/mol
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00150598
CAS Number: 149-44-0
EINECS: 205-739-4
Merck Index: 14,8620

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR