3-(4-Morpholinyl)phenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15707-03
A15707-06
A15707-14
CAAAAA15707-03EA
67.62
CAD
CAAAAA15707-03
CAAAAA15707-06
CAAAAA15707-14
3-(4-Morpholinyl)phenol 98%
3-(4-Morpholinyl)phenol
Purity: 98%
CAS No.: 27292-49-5
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point: 127-132°
Beilstein Reference: 149989
Formula:
C₁₀H₁₃NO₂ Melting Pt: 127…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051675 CAS Number: 27292-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...