N,N-Diethylcarbamoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A15701-14 A15701-22
CAAAAA15701-14EA 39.18 CAD
CAAAAA15701-14 CAAAA15701-22
N,N-Diethylcarbamoyl chloride 98%
N,N-Diethylcarbamoyl chloride

Purity: 98% CAS No.: 88-10-8 Molecular Formula: C5H10ClNO Formula Weight: 135.59 Flash Point: 76°(168°F) Refractive Index: 1.4510 Melting Point: -32° Boiling Point: 186° Beilstein Reference: 506687 UN No.: 3265 MDL No.: MFCD00000636

Formula: C₅H₁₀ClNO
Boiling Pt: 186 °C
Melting Pt: –32 °C
Density: 1.070
Flash Pt: 76 °C(168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000636
CAS Number: 88-10-8
EINECS: 201-798-5
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR