Benzyl chloroformate 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃) ∼0.1%
A15682-30
A15682-0B
A15682-18
CAAAAA15682-30EA
350.76
CAD
CAAAAA15682-30
CAAAA15682-0B
CAAAA15682-18
Benzyl chloroformate 95% stabilized
Benzyl chloroformate
Purity: 95%
CAS No.: 501-53-1
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Storage Temperature: -30°C to -10°C
Flash Point: 125°(257°F)
Refractive Index: 1.5170
Odor: Acrid
Melting Point: -20°
Boiling Point: 178-180°
Merck Reference: 14,1801
UN No.: 1739
MDL No.: MFCD00000640
Formula:
C₈H₇ClO₂ Boiling Pt: 178…180 °C Melting Pt: –20 °C Density: 1.195 Flash Pt: 125 °C (257 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000640 CAS Number: 501-53-1 EINECS: 207-925-0 UN: 1739 ADR: 8,I Merck Index: 14,1801 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...