D-(-)-α-Phenylglycine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-(-)-2-Phenylglycine, H-D-Phg-OH, (R)-(-)-α-Aminophenylacetic acid
A15669-14
A15669-22
A15669-36
CAAAA15669-14EA
43.61
CAD
CAAAA15669-14
CAAAA15669-22
CAAAA15669-36
D-(-)-α-Phenylglycine 99%
D(-)-α-Phenylglycine
Purity: 99%
CAS No.: 875-74-1
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 277° dec.
Beilstein Reference: 2208676
Merck Reference: 14,7291
Optical Rotation: -154° (c=1 in 1N HCl)
MDL No.: MFCD00008061
Formula:
C₈H₉NO₂ Melting Pt: 277 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008061 CAS Number: 875-74-1 EINECS: 212-876-3 Merck Index: 14,7291 |
Specification Test Results
Optical rotation | -154° (c=1 in 1N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...