Ethyl-4-chloroacetoacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15641-0B
A15641-18
A15641-30
CAAAA15641-0BEA
587.76
CAD
CAAAA15641-0B
CAAAA15641-18
CAAAA15641-30
Ethyl-4-chloroacetoacetate 97%
Ethyl 4-chloroacetoacetate
Purity: 97%
CAS No.: 638-07-3
Molecular Formula: C6H9ClO3
Formula Weight: 164.59
Flash Point: 92°(198°F)
Refractive Index: 1.4520
Melting Point: -8°
Boiling Point: 220°
Beilstein Reference: 1761275
UN No.: 2922
MDL No.: MFCD00000939
Formula:
C₆H₉ClO₃ Boiling Pt: 220 °C Melting Pt: –8 °C Density: 1.212 Flash Pt: 92 °C(198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000939 CAS Number: 638-07-3 EINECS: 211-317-0 UN: 2927 ADR: "6.1",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...