Chloro(dimethyl)(phenyl)silane 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorodimethylphenylsilane
A15638-06
A15638-14
A15638-22
CAAAA15638-06EA
72.21
CAD
CAAAA15638-06
CAAAA15638-14
CAAAA15638-22
Chloro(dimethyl)(phenyl)silane 97+%
Chloro(dimethyl)(phenyl)silane
Fieser: 2,278 7,133 8,196 10,162 11,209 12,210 16,146 20,304
Purity: 97+%
CAS No.: 768-33-2
Molecular Formula: C8H11ClSi
Formula Weight: 170.72
Flash Point: 72°(161°F)
Refractive Index: 1.5090
Boiling Point: 192-193°
Beilstein Reference: 606292
UN No.: 2987
MDL No.: MFCD00000499
Formula:
C₈H₁₁ClSi Boiling Pt: 192…193 °C Density: 1.032 Flash Pt: 72 °C(161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000499 CAS Number: 768-33-2 EINECS: 212-193-0 UN: 2987 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...