Lead tetraacetate 96% (dry weight) stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Moistured with glacial acetic acid 5-10%
A15551-0B
A15551-22
A15551-30
CAAAAA15551-0BEA
654.12
CAD
CAAAAA15551-0B
CAAAAA15551-22
CAAAAA15551-30
CAAAAA15551-36
Lead tetraacetate 96% (dry weight) stabilized
Lead tetraacetate
Fieser: 1,537 9,265 12,270 13,155 14,188 16,193 19,184 20,209 21,245
Purity: 96%
CAS No.: 546-67-8
Molecular Formula: C8H12O8Pb
Formula Weight: 443.37
Melting Point: 175°
Beilstein Reference: 3595640
Merck Reference: 14,5423
UN No.: 1616
MDL No.: MFCD00008693
Formula:
(H₃CCOO)₄Pb Melting Pt: 175 °C Density: 2.28 Storage Temperature: Ambient |
MDL Number:
MFCD00008693 CAS Number: 546-67-8 EINECS: 208-908-0 UN: 1616 ADR: 6.1,III Merck Index: 14,5423 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...