Saccharin sodium hydrate 99%

Supplier: Thermo Scientific Chemicals

A15530-0B A15530-0I A15530-30
CAAAAA15530-0BEA 183.28 CAD
CAAAAA15530-0B CAAAAA15530-0I CAAAAA15530-30
Saccharin sodium hydrate 99%
Saccharin sodium hydrate

Purity: 99% CAS No.: 82385-42-0 Molecular Formula: C7H4NNaO3S.xH2O Formula Weight: 205.16(anhy) Melting Point: 300° Beilstein Reference: 3599229 Merck Reference: 14,8312 MDL No.: MFCD00151213

Formula: C₇H₅NNaO₃S·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149605
CAS Number: 82385-42-0
EINECS: 204-886-1
Merck Index: 14,8312

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR