Aminoacetaldehyde dimethyl acetal 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Dimethoxyethanamine
A15498-14
A15498-22
A15498-36
CAAAA15498-14EA
87.69
CAD
CAAAA15498-14
CAAAA15498-22
CAAAA15498-36
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal
Purity: 99%
CAS No.: 22483-09-6
Molecular Formula: H2NCH2CH(OCH3)2
Formula Weight: 105.14
Flash Point: 41°(106°F)
Refractive Index: 1.4170
Boiling Point: 135-138°
Beilstein Reference: 741868
UN No.: 2733
MDL No.: MFCD00008135
Formula:
C₄H₁₁NO₂ Boiling Pt: 135…138 °C Density: 0.973 Flash Pt: 41 °C(106 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008135 CAS Number: 22483-09-6 EINECS: 245-026-5 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...