Ethyl propionylacetate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15464-22
A15464-06
A15464-14
CAAAAA15464-22EA
1606.86
CAD
CAAAAA15464-22
CAAAA15464-06
CAAAA15464-14
Ethyl propionylacetate 95%
Ethyl propionylacetate
Purity: 95%
CAS No.: 4949-44-4
Molecular Formula: CH3CH2COCH2CO2CH2CH3
Formula Weight: 144.17
Flash Point: 77°(170°F)
Refractive Index: 1.4220
Boiling Point: 190-192°
Beilstein Reference: 970270
MDL No.: MFCD00009317
Formula:
C₇H₁₂O₃ Boiling Pt: 190…192 °C Density: 1.012 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009317 CAS Number: 4949-44-4 EINECS: 225-593-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...