(2-Naphthoxy)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A15412-22 A15412-36
CAAAA15412-0EEA 832.8 CAD
CAAAA15412-0E CAAAA15412-22 CAAAA15412-36
(2-Naphthoxy)acetic acid 97%
(2-Naphthoxy)acetic acid

Purity: 97% CAS No.: 120-23-0 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 155-157° Beilstein Reference: 1074148 Merck Reference: 14,6397 MDL No.: MFCD00004066

Formula: C₁₂H₁₀O₃
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00004066
CAS Number: 120-23-0
EINECS: 204-380-0
Merck Index: 14,6397

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR