Aluminon
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15401-09
A15401-18
A15401-30
CAAAAA15401-09EA
55.93
CAD
CAAAAA15401-09
CAAAAA15401-18
CAAAAA15401-30
Aluminon
Aluminon
CAS No.: 569-58-4
Molecular Formula: C22H23N3O9
Formula Weight: 473.44
Odor: Odorless
pH: 5.3
Melting Point: 220-225° dec.
MDL No.: MFCD00040925
Formula:
C₂₂H₂₃N₃O₉ Melting Pt: 220…225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00040925 CAS Number: 569-58-4 EINECS: 209-319-1 Merck Index: 14,322 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...