2,5-Diphenyl-p-benzoquinone 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Diphenylquinone, [1,1:4,1-Terphenyl]-2,5-dione

A15369-06 A15369-14
CAAAA15369-06EA 71.42 CAD
CAAAA15369-06 CAAAA15369-14
2,5-Diphenyl-p-benzoquinone 96%
2,5-Diphenyl-p-benzoquinone

Purity: 96% CAS No.: 844-51-9 Molecular Formula: C18H12O2 Formula Weight: 260.30 Melting Point: 215-219° Beilstein Reference: 2051812

Formula: C₁₈H₁₂O₂
MW: 260.29 g/mol
Melting Pt: 215…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00001600
CAS Number: 844-51-9
EINECS: 212-680-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR