Creatine phosphate disodium salt tetrahydrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

A15362-03 A15362-06 A15362-14
CAAAA15362-03EA 65.41 CAD
CAAAA15362-03 CAAAA15362-06 CAAAA15362-14
Creatine phosphate disodium salt tetrahydrate 98+%
Creatine phosphate disodium salt tetrahydrate

Substrate for creatine kinase determinations.


Purity: 98+% CAS No.: 71519-72-7 Molecular Formula: C4H8N3Na2O5P.4H2O Formula Weight: 327.14 (255.07anhy) Storage Temperature: 2°C to 8°C


Substrate for creatine kinase determinations

Formula: Na₂O₃PNHC(=NH)N(CH₃)CH₂COOH·4H₂O
MW: 327.14 g/mol (255.07 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150192
CAS Number: 71519-72-7
EINECS: 213-074-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR