(4-Hydroxyphenyl)acetamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15266-09
A15266-18
CAAAA15266-09EA
46.93
CAD
CAAAA15266-09
CAAAA15266-18
CAAAA15266-30
(4-Hydroxyphenyl)acetamide 99%
(4-Hydroxyphenyl)acetamide
Purity: 99%
CAS No.: 17194-82-0
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 173-175°
Beilstein Reference: 2085984
Formula:
C₈H₉NO₂ Melting Pt: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017145 CAS Number: 17194-82-0 EINECS: 241-235-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...