Uridine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
A15227-06
A15227-14
A15227-22
CAAAA15227-06EA
63.2
CAD
CAAAA15227-06
CAAAA15227-14
CAAAA15227-22
Uridine 99%
Uridine
Purity: 99%
CAS No.: 58-96-8
Molecular Formula: C9H12N2O6
Formula Weight: 244.20
Melting Point: 166-169°
Beilstein Reference: 754904
Merck Reference: 14,9877
Optical Rotation: +9.5° (c=2 in water)
MDL No.: MFCD00006526
Formula:
C₉H₁₂N₂O₆ Melting Pt: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006526 CAS Number: 58-96-8 EINECS: 200-407-5 Merck Index: 14,9877 |
Specification Test Results
Optical rotation | +9.5° (c=2 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...