(±)-Tetrahydrofurfuryl bromide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15192-09
A15192-18
CAAAA15192-09EA
111.86
CAD
CAAAA15192-09
CAAAA15192-18
CAAAA15192-30
(±)-Tetrahydrofurfuryl bromide 95%
(±)-Tetrahydrofurfuryl bromide
Purity: 95%
CAS No.: 1192-30-9
Molecular Formula: C5H9BrO
Formula Weight: 165.04
Storage Temperature: 2°C to 8°C
Flash Point: 61°(142°F)
Refractive Index: 1.4880
Boiling Point: 169-171°
Beilstein Reference: 102717
MDL No.: MFCD00016894
Formula:
C₅H₉BrO Boiling Pt: 169…171 °C Density: 1.452 Flash Pt: 61 °C(142 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00016894 CAS Number: 1192-30-9 EINECS: 214-750-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...