Diphenyliodonium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15149-09
A15149-18
A15149-30
CAAAAA15149-09EA
159.58
CAD
CAAAAA15149-09
CAAAAA15149-18
CAAAAA15149-30
Diphenyliodonium chloride 98+%
Diphenyliodonium chloride
Fieser: 1,341 2,178 15,150
Purity: 98+%
CAS No.: 1483-72-3
Molecular Formula: C12H10ClI
Formula Weight: 316.57
Melting Point: ca 230° dec.
Beilstein Reference: 3574977
Merck Reference: 14,7464
UN No.: 2811
MDL No.: MFCD00011909
Formula:
C₁₂H₁₀ClI Melting Pt: ca 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011909 CAS Number: 1483-72-3 EINECS: 216-049-8 UN: 2811 ADR: "6.1",III Merck Index: 14,7464 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...