(R)-(-)-Thiazolidine-4-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid

A15033-09 A15033-18 A15033-30
CAAAA15033-09EA 45.98 CAD
CAAAA15033-09 CAAAA15033-18 CAAAA15033-30
(R)-(-)-Thiazolidine-4-carboxylic acid 98%
(R)-(-)-Thiazolidine-4-carboxylic acid

Purity: 98% CAS No.: 34592-47-7 Molecular Formula: C4H7NO2S Formula Weight: 133.17 Melting Point: 192-200° dec. Beilstein Reference: 81065 Merck Reference: 14,9445 Optical Rotation: -141° (c=1.3 in water) UN No.: 2811 MDL No.: MFCD00005212

Formula: C₄H₇NO₂S
MW: 133.17 g/mol
Melting Pt: 192…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005212
CAS Number: 34592-47-7
EINECS: 252-106-3
ADR: 6.1,III
Merck Index: 14,9445

Order Now

Specification Test Results

Optical rotation -141° (c=1.3 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR