Dimethyl-3-oxoglutarate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

A14969-14 A14969-22 A14969-36
CAAAA14969-14EA 60.99 CAD
CAAAA14969-14 CAAAA14969-22 CAAAA14969-36
Dimethyl-3-oxoglutarate 97%
Dimethyl 3-oxoglutarate

Purity: 97% CAS No.: 1830-54-2 Molecular Formula: C7H10O5 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4440 Odor: Ester-like Melting Point: 16-17° Boiling Point: 237-241° Beilstein Reference: 1366042 MDL No.: MFCD00008462

Formula: C₇H₁₀O₅
Boiling Pt: 237…241 °C
Melting Pt: 16…17 °C
Density: 1.185
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR