(±)-Camphorquinone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

A14967-06 A14967-14 A14967-22
CAAAA14967-06EA 123.71 CAD
CAAAA14967-06 CAAAA14967-14 CAAAA14967-22
(±)-Camphorquinone 99%
(±)-Camphorquinone

Purity: 99% CAS No.: 10373-78-1 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 198-202° Beilstein Reference: 1909463

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00064160
CAS Number: 10373-78-1
EINECS: 233-814-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR