Methylenediphosphonic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14803-06
A14803-14
CAAAA14803-06EA
455.04
CAD
CAAAA14803-06
CAAAA14803-14
Methylenediphosphonic acid 99+%
Methylenediphosphonic acid
Purity: 99+%
CAS No.: 1984-15-2
Molecular Formula: CH6O6P2
Formula Weight: 176.00
Odor: Odorless
Melting Point: 202-204°
Beilstein Reference: 1708494
Merck Reference: 14,5792
UN No.: 3261
MDL No.: MFCD00002139
Formula:
CH₆O₆P₂ Melting Pt: 202…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002139 CAS Number: 1984-15-2 EINECS: 217-851-0 UN: 3261 ADR: 8,III Merck Index: 14,5792 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...