Ethyl oxalyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14653-14
A14653-22
A14653-36
CAAAA14653-14EA
73
CAD
CAAAA14653-14
CAAAA14653-22
CAAAA14653-36
Ethyl oxalyl chloride 98%
Ethyl oxalyl chloride
Purity: 98%
CAS No.: 4755-77-5
Molecular Formula: C4ClH5O3
Formula Weight: 136.53
Flash Point: 41°(105°F)
Refractive Index: 1.4160
Boiling Point: 134-135°
Beilstein Reference: 506725
UN No.: 2920
MDL No.: MFCD00000706
Formula:
C₄H₅ClO₃ Boiling Pt: 134…135 °C Density: 1.226 Flash Pt: 41 °C(105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000706 CAS Number: 4755-77-5 EINECS: 225-285-0 UN: 2920 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...