Isatoic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Benzo[d][1,3]oxazine-2,4-dione
A14603-0E
A14603-22
A14603-36
CAAAA14603-0EEA
663.6
CAD
CAAAA14603-0E
CAAAA14603-22
CAAAA14603-36
Isatoic anhydride 97%
Isatoic anhydride
Purity: 97%
CAS No.: 118-48-9
Molecular Formula: C8H5NO3
Formula Weight: 163.13
Flash Point: 308°(586°F)
Melting Point: ca 237° dec.
Beilstein Reference: 136786
MDL No.: MFCD00006700
Formula:
C₈H₅NO₃ Melting Pt: ca 237 °C (dec.) Density: 1.520 Flash Pt: 308 °C(586 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006700 CAS Number: 118-48-9 EINECS: 204-255-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...