Methyl carbazate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14548-0E
A14548-22
CAAAA14548-0EEA
1203.96
CAD
CAAAA14548-0E
CAAAA14548-22
Methyl carbazate 98%
Methyl carbazate
Fieser: 4,333 7,235
Purity: 98%
CAS No.: 6294-89-9
Molecular Formula: C2H6N2O2
Formula Weight: 90.08
Odor: Weak
Melting Point: 70-73°
Beilstein Reference: 1679395
UN No.: 2811
MDL No.: MFCD00007594
Formula:
C₂H₆N₂O₂ Melting Pt: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007594 CAS Number: 6294-89-9 EINECS: 228-560-3 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...