Riboflavine-5'-phosphate sodium salt dihydrate

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate

A14545-09 A14545-18
CAAAA14545-09EA 79.79 CAD
CAAAA14545-09 CAAAA14545-18
Riboflavine-5'-phosphate sodium salt dihydrate
Riboflavine-5'-phosphate sodium salt dihydrate

Photoinitiator of polymerization for PAGE gels


CAS No.: 6184-17-4 Molecular Formula: C17H20N4NaO9P.2H2O Formula Weight: 514.36 (478.34anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 4106529 Merck Reference: 14,8201 MDL No.: MFCD00150993

Formula: C₁₇H₂₀N₄NaO₉P·2H₂O
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00150993
CAS Number: 6184-17-4
EINECS: 204-988-6
Merck Index: 14,8201

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR