Allyltriphenylphosphonium bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14490-18
A14490-30
CAAAA14490-18EA
83.11
CAD
CAAAA14490-18
CAAAA14490-30
Allyltriphenylphosphonium bromide 99%
Allyltriphenylphosphonium bromide
Purity: 99%
CAS No.: 1560-54-9
Molecular Formula: C21H20BrP
Formula Weight: 383.27
Melting Point: 222-225°
Beilstein Reference: 3579053
Formula:
C₂₁H₂₀BrP Melting Pt: 222…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011808 CAS Number: 1560-54-9 EINECS: 216-332-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...