Sodium-3-(trimethylsilyl)propionate-[D₄] (98% D)

Supplier: Thermo Scientific Chemicals
Warning

A14489-MD A14489-03 A14489-06
CAAAAA14489-MDEA 134.93 CAD
CAAAAA14489-MD CAAAA14489-03 CAAAA14489-06
Sodium-3-(trimethylsilyl)propionate-[D₄] (98% D)
Sodium 3-(trimethylsilyl)propionate-[D4]

Purity: 98+ atom %D CAS No.: 24493-21-8 Molecular Formula: C6H9D4NaO2Si Formula Weight: 172.27 Melting Point: 300° Beilstein Reference: 4046239

Formula: C₆H₉D₄NaO₂Si
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002762
CAS Number: 24493-21-8
EINECS: 246-286-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR