Inosine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

A14459-06 A14459-14 A14459-22
CAAAA14459-06EA 49.77 CAD
CAAAA14459-06 CAAAA14459-14 CAAAA14459-22
Inosine 98+%
Inosine

Uses: Suppresses the increase of glucose and insulin used in blood


Applications: Suppresses the increase of glucose and insulin used in blood.

Formula: C₁₀H₁₂N₄O₅
MW: 268.23 g/mol
Melting Pt: 212…215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066770
CAS Number: 58-63-9
EINECS: 200-390-4
Merck Index: 14,4975

Order Now

Specification Test Results

Optical rotation -52° (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR