1-Acetyl-2-phenylhydrazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14446-22
A14446-36
CAAAA14446-22EA
159.58
CAD
CAAAA14446-22
CAAAA14446-36
1-Acetyl-2-phenylhydrazine 98%
1-Acetyl-2-phenylhydrazine
Purity: 98%
CAS No.: 114-83-0
Molecular Formula: C6H5NHNHCOCH3
Formula Weight: 150.18
Melting Point: 128-131°
Beilstein Reference: 742880
UN No.: 2811
MDL No.: MFCD00008672
Formula:
C₈H₁₀N₂O Melting Pt: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008672 CAS Number: 114-83-0 EINECS: 204-055-3 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...