Triethyloxonium tetrafluoroborate 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with diethyl ether 3-5%

A14420-14 A14420-22 A14420-36
CAAAA14420-14EA 169.06 CAD
CAAAA14420-14 CAAAA14420-22 CAAAA14420-36
Triethyloxonium tetrafluoroborate 95% stabilized
Triethyloxonium tetrafluoroborate

Fieser: 1,1210 4,527 5,691 6,611 7,386 8,500 9,482 10,417 15,337


Purity: 95% CAS No.: 368-39-8 Molecular Formula: C6H15BF4O Formula Weight: 189.99 Storage Temperature: 2°C to 8°C Melting Point: 84-89° Beilstein Reference: 3598090 UN No.: 2925 MDL No.: MFCD00044423

Formula: C₆H₁₅BF₄O
Melting Pt: 84…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044423
CAS Number: 368-39-8
EINECS: 206-705-1
UN: 2925
ADR: "4.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR