2-Hydroxy-3-methoxy-5-nitrobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14360-03
A14360-06
A14360-14
CAAAA14360-03EA
54.19
CAD
CAAAA14360-03
CAAAA14360-06
CAAAA14360-14
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde 98%
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde
Purity: 98%
CAS No.: 17028-61-4
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 138-142°
Beilstein Reference: 613271
Formula:
C₈H₇NO₅ MW: 197.15 g/mol Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017033 CAS Number: 17028-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...