Ethyl mercaptoacetate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-mercaptoacetate, Ethyl thioglycolate

A14321-06 A14321-22 A14321-36
CAAAAA14321-06EA 44.4 CAD
CAAAAA14321-06 CAAAA14321-22 CAAAA14321-36
Ethyl mercaptoacetate 98+%
Ethyl mercaptoacetate

Purity: 98+% CAS No.: 623-51-8 Molecular Formula: C4H8O2S Formula Weight: 120.17 Flash Point: 48°(118°F) Refractive Index: 1.4580 Odor: Stench Melting Point: -80° Boiling Point: 156-159° Beilstein Reference: 605581 UN No.: 1228 MDL No.: MFCD00004874

Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 156…159 °C
Melting Pt: –80 °C
Density: 1.096
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004874
CAS Number: 623-51-8
EINECS: 210-800-3
UN: 1228
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR