2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94% cont. up to ca 6% acetone
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,6-Trimethyl-1,3-dioxine-4-one
A14254-14
A14254-22
A14254-36
CAAAA14254-14EA
53.4
CAD
CAAAA14254-14
CAAAA14254-22
CAAAA14254-36
2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94% cont. up to ca 6% acetone
2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Fieser: 10,424
Purity: 94%
CAS No.: 5394-63-8
Molecular Formula: C7H10O3
Formula Weight: 142.15
Storage Temperature: 2°C to 8°C
Flash Point: 14°(57°F)
Refractive Index: 1.4610
Melting Point: 7°
Boiling Point: 65-67°/2mm
Beilstein Reference: 2408
UN No.: 1993
MDL No.: MFCD00040468
Formula:
C₇H₁₀O₃ MW: 142.15 g/mol Boiling Pt: 65…67 °C/2 mm Melting Pt: 7 °C Density: 1.094 Flash Pt: 14 °C(57 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00040468 CAS Number: 5394-63-8 EINECS: 226-403-3 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...