2-(Chloromethyl)quinoline hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14102-03
A14102-06
CAAAA14102-03EA
51.35
CAD
CAAAA14102-03
CAAAA14102-06
CAAAA14102-14
2-(Chloromethyl)quinoline hydrochloride 97%
2-(Chloromethyl)quinoline hydrochloride
Purity: 97%
CAS No.: 3747-74-8
Molecular Formula: C10H8ClN.HCl
Formula Weight: 214.10
Melting Point: 184-190°
Beilstein Reference: 3565832
Formula:
C₁₀H₉Cl₂N Melting Pt: 184…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012734 CAS Number: 3747-74-8 EINECS: 223-145-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...